2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-tert-Butyl-4-methylthiazole-5-carboxylic acid
H32501.MD
H32501.03
ALFAH32501.MDEA
0
SGD
ALFAH32501.MD
ALFAH32501.03
2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid ≥98%
2-tert-Butyl-4-methyl-5-thiazolecarboxylic acid
Purity: 98%
Molecular Formula: C9H13NO2S
Formula Weight: 199.27
Melting Point: 154-157°
MDL No.: MFCD11207787
Formula:
C₉H₁₃NO₂S MW: 199,27 g/mol Melting Pt: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD11207787 CAS Number: 427896-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...