Imidazo[1,2-a]pyridine-6-carboxylic acid 95%

Supplier: Thermo Fisher Scientific
Warning

H33128.03 H33128.06
ALFAH33128.03EA 0 SGD
ALFAH33128.03 ALFAH33128.06
Imidazo[1,2-a]pyridine-6-carboxylic acid 95%
Imidazo[1,2-a]pyridine-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 244…247 °C
Storage Temperature: Ambient
MDL Number: MFCD07021498
CAS Number: 139022-25-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR