2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%
Supplier: Thermo Scientific
L19575.MC
L19575.03
ALFAL19575.MCEA
0
SGD
ALFAL19575.MC
ALFAL19575.03
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester
Purity: 98%
CAS No.: 501014-45-5
Molecular Formula: C18H21BN2O2
Formula Weight: 308.19
Melting Point: 244-248°
MDL No.: MFCD03788733
Formula:
C₁₈H₂₁BN₂O₂ MW: 308,19 g/mol Melting Pt: 244…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788733 CAS Number: 501014-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...