2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
Supplier: Thermo Scientific
|
Danger
|
H32375.03
H32375.06
ALFAH32375.03EA
0
SGD
ALFAH32375.03
ALFAH32375.06
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine
Purity: 97%
CAS No.: 1023-01-4
Molecular Formula: C14H11BrN2
Formula Weight: 287.16
Melting Point: 214-218°
MDL No.: MFCD00444784
Formula:
C₁₄H₁₁BrN₂ MW: 287,16 g/mol Melting Pt: 214…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00444784 CAS Number: 1023-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...