Methyl quinoxaline-6-carboxylate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 6-Quinoxalinecarboxylate
A16689.06
A16689.03
ALFAA16689.06EA
0
SGD
ALFAA16689.06
ALFAA16689.03
Methyl quinoxaline-6-carboxylate ≥98%
Methyl quinoxaline-6-carboxylate
Purity: 98%
CAS No.: 23088-23-5
Molecular Formula: C10H8N2O2
Formula Weight: 188.19
Melting Point: 96-99°
MDL No.: MFCD00102618
Formula:
C₁₀H₈N₂O₂ MW: 188,19 g/mol Melting Pt: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102618 CAS Number: 23088-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...