1H-Indazole-4-carboxylic acid ≥97%

Supplier: Thermo Scientific
Warning

H34115.MD H34115.03 H34115.06
ALFAH34115.MDEA 0 SGD
ALFAH34115.MD ALFAH34115.03 ALFAH34115.06
1H-Indazole-4-carboxylic acid ≥97%
1H-Indazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162,15 g/mol
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03840644
CAS Number: 677306-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR