Ethyl-5-methylisoxazole-4-carboxylate 96%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Ethyl 5-methylisoxazole-4-carboxylate
L18647.09
L18647.04
ALFAL18647.09EA
0
SGD
ALFAL18647.09
ALFAL18647.04
Ethyl-5-methylisoxazole-4-carboxylate 96%
Ethyl 5-methylisoxazole-4-carboxylate
Purity: 96%
CAS No.: 51135-73-0
Molecular Formula: C7H9NO3
Formula Weight: 155.15
Refractive Index: 1.4597
Boiling Point: 95-97°/15mm
MDL No.: MFCD01631119
Formula:
C₇H₉NO₃ MW: 155,15 g/mol Boiling Pt: 95…97 °C (15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD01631119 CAS Number: 51135-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...