2-(4-Chlorophenyl)ethylboronic acid pinacol ester ≥98%

Supplier: Thermo Scientific

Synonyms: 2-(4-Chlorophenethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H30303.03 H30303.06
ALFAH30303.03EA 0 SGD
ALFAH30303.03 ALFAH30303.06
2-(4-Chlorophenyl)ethylboronic acid pinacol ester ≥98%
2-(4-Chlorophenyl)ethylboronic acid pinacol ester

Purity: 98% CAS No.: 444094-88-6 Molecular Formula: C14H20BClO2 Formula Weight: 266.58 Melting Point: 34-38° MDL No.: MFCD12546184

Formula: C₁₄H₂₀BClO₂
MW: 266,58 g/mol
Melting Pt: 34…38 °C
Storage Temperature: Ambient
MDL Number: MFCD12546184
CAS Number: 444094-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR