4-(Cyclopentylcarbamoyl)phenylboronic acid pinacol ester 95%

Supplier: Thermo Scientific

H52269.03 H52269.06
ALFAH52269.03EA 71.2 SGD
ALFAH52269.03 ALFAH52269.06
4-(Cyclopentylcarbamoyl)phenylboronic acid pinacol ester 95%
4-(Cyclopentylcarbamoyl)phenylboronic acid pinacol ester

Purity: 95% CAS No.: 933987-10-1 Molecular Formula: C18H26BNO3 Formula Weight: 315.22 MDL No.: MFCD16660857

Formula: C₁₈H₂₆BNO₃
MW: 315,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660857
CAS Number: 933987-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR