3-(Dimethylcarbamoyl)phenylboronic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)benzeneboronic acid, (3-(Dimethylcarbamoyl)phenyl)boronic acid

H52545.MD H52545.03
ALFAH52545.MDEA 0 SGD
ALFAH52545.MD ALFAH52545.03
3-(Dimethylcarbamoyl)phenylboronic acid 96%
3-(Dimethylcarbamoyl)phenylboronic acid

Purity: 96% CAS No.: 373384-14-6 Molecular Formula: C9H12BNO3 Formula Weight: 193.01 MDL No.: MFCD03412066

Formula: C₉H₁₂BNO₃
MW: 193,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412066
CAS Number: 373384-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR