2-Bromo-N-methylbenzamide ≥99%

Supplier: Thermo Scientific
Warning

H50583.03 H50583.06
ALFAH50583.03EA 262 SGD
ALFAH50583.03 ALFAH50583.06
2-Bromo-N-methylbenzamide ≥99%
2-Bromo-N-methylbenzamide

Purity: 99% CAS No.: 61436-88-2 Molecular Formula: C8H8BrNO Formula Weight: 214.06 MDL No.: MFCD00099250

Formula: C₈H₈BrNO
MW: 214,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00099250
CAS Number: 61436-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR