2-Amino-4-chlorothiobenzamide 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Amino-4-chlorobenzothioamide
H52249.03
H52249.06
ALFAH52249.03EA
215
SGD
ALFAH52249.03
ALFAH52249.06
2-Amino-4-chlorothiobenzamide 97%
2-Amino-4-chlorothiobenzamide
Purity: 97%
CAS No.: 2586-93-8
Molecular Formula: C7H7ClN2S
Formula Weight: 186.66
MDL No.: MFCD16658869
Formula:
C₇H₇ClN₂S MW: 186,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16658869 CAS Number: 2586-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...