3,5-Dinitro-4-methylphenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methyl-3,5-dinitrobenzeneboronic acid

H25773.03 H25773.06 H25773.14
ALFAH25773.03EA 0 SGD
ALFAH25773.03 ALFAH25773.06 ALFAH25773.14
3,5-Dinitro-4-methylphenylboronic acid ≥97%
3,5-Dinitro-4-methylphenylboronic acid

Purity: 97% CAS No.: 28249-49-2 Molecular Formula: C7H7BN2O6 Formula Weight: 225.95 Melting Point: ca 280° dec. MDL No.: MFCD08060935

Formula: C₇H₇BN₂O₆
MW: 225,95 g/mol
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08060935
CAS Number: 28249-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR