Methyl-3,5-dibromo-2,4-dihydroxy-6-methylbenzoate ≥99%

Supplier: Thermo Scientific
Warning

L12098.03 L12098.06
ALFAL12098.03EA 0 SGD
ALFAL12098.03 ALFAL12098.06
Methyl-3,5-dibromo-2,4-dihydroxy-6-methylbenzoate ≥99%
Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate

Purity: 99% CAS No.: 715-33-3 Molecular Formula: C9H8Br2O4 Formula Weight: 339.98 Melting Point: 104-107° Beilstein Reference: 2116569

Formula: C₉H₈Br₂O₄
MW: 339,98 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00082715
CAS Number: 715-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR