4-Chloro-3-(trifluoromethyl)thiobenzamide ≥97%

Supplier: Thermo Scientific
Danger

H33536.MD H33536.03
ALFAH33536.MDEA 0 SGD
ALFAH33536.MD ALFAH33536.03
4-Chloro-3-(trifluoromethyl)thiobenzamide ≥97%
4-Chloro-3-(trifluoromethyl)thiobenzamide

Purity: 97% CAS No.: 1315339-86-6 Molecular Formula: C8H5ClF3NS Formula Weight: 239.65 Melting Point: 68-71° MDL No.: MFCD09757541

Formula: C₈H₅ClF₃NS
MW: 239,65 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD09757541
CAS Number: 1315339-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR