Methyl-3,5-bis(trifluoromethyl)benzoate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3,5-bis(trifluoromethyl)benzoate

B20990.03 B20990.06
ALFAB20990.03EA 0 SGD
ALFAB20990.03 ALFAB20990.06
Methyl-3,5-bis(trifluoromethyl)benzoate ≥98%
Methyl 3,5-bis(trifluoromethyl)benzoate

Purity: 98% CAS No.: 26107-80-2 Molecular Formula: C10H6F6O2 Formula Weight: 272.15 Boiling Point: 85-87°/15mm MDL No.: MFCD00052317

Formula: C₁₀H₆F₆O₂
MW: 272,15 g/mol
Boiling Pt: 85…87 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052317
CAS Number: 26107-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR