5-Iodo-2-(4-morpholino)pyridine 96%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(5-Iodo-2-pyridinyl)morpholine, 4-(5-Iodo-2-pyridyl)morpholine

H50142.03 H50142.MD
ALFAH50142.03EA 0 SGD
ALFAH50142.03 ALFAH50142.MD
5-Iodo-2-(4-morpholino)pyridine 96%
5-Iodo-2-(4-morpholino)pyridine

Purity: 96% CAS No.: 470463-42-4 Molecular Formula: C9H11IN2O Formula Weight: 290.11 Melting Point: 131-135° MDL No.: MFCD04039866

Formula: C₉H₁₁IN₂O
MW: 290,11 g/mol
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD04039866
CAS Number: 470463-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR