4-(Benzyloxycarbamoyl)phenylboronic acid pinacol ester 95%
Supplier: Thermo Scientific
H52282.03
H52282.06
ALFAH52282.03EA
71.6
SGD
ALFAH52282.03
ALFAH52282.06
4-(Benzyloxycarbamoyl)phenylboronic acid pinacol ester 95%
4-(Benzyloxycarbamoyl)phenylboronic acid pinacol ester
Purity: 95%
Molecular Formula: C20H24BNO4
Formula Weight: 353.22
MDL No.: MFCD18827484
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...