4-Benzyloxy-2-fluorophenylboronic acid 96%

Supplier: Thermo Scientific
Warning

L18521.06 L18521.MD
ALFAL18521.06EA 0 SGD
ALFAL18521.06 ALFAL18521.MD
4-Benzyloxy-2-fluorophenylboronic acid 96%
4-Benzyloxy-2-fluorophenylboronic acid

Purity: 96% CAS No.: 166744-78-1 Molecular Formula: C13H12BFO3 Formula Weight: 246.04 MDL No.: MFCD02093064

Formula: C₁₃H₁₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD02093064
CAS Number: 166744-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR