Isoquinoline-3-carbonitrile ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Isoquinolinecarbonitrile
L19426.03
L19426.MD
ALFAL19426.03EA
0
SGD
ALFAL19426.03
ALFAL19426.MD
Isoquinoline-3-carbonitrile ≥99%
Isoquinoline-3-carbonitrile
Purity: 99%
CAS No.: 26947-41-1
Molecular Formula: C10H6N2
Formula Weight: 154.17
Melting Point: 125-128°
Beilstein Reference: 114887
Formula:
C₁₀H₆N₂ MW: 154,17 g/mol Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075136 CAS Number: 26947-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...