3-Chloro-2-methylbenzonitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
L11048.03
L11048.06
ALFAL11048.03EA
89.4
SGD
ALFAL11048.03
ALFAL11048.06
3-Chloro-2-methylbenzonitrile ≥97%
3-Chloro-2-methylbenzonitrile
Purity: 97%
CAS No.: 54454-12-5
Molecular Formula: C8H6ClN
Formula Weight: 151.60
Refractive Index: 1.5530
Melting Point: 20-23°
Boiling Point: 120°/22mm
Beilstein Reference: 2613370
UN No.: 3439
MDL No.: MFCD00045598
Formula:
C₈H₆ClN MW: 151,60 g/mol Boiling Pt: 120 °C (22 mmHg) Melting Pt: 20…23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045598 CAS Number: 54454-12-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...