3,4-Difluoro-2-methylbenzonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
H31548.03
H31548.06
ALFAH31548.03EA
0
SGD
ALFAH31548.03
ALFAH31548.06
3,4-Difluoro-2-methylbenzonitrile ≥98%
3,4-Difluoro-2-methylbenzonitrile
Purity: 98%
CAS No.: 847502-83-4
Molecular Formula: C8H5F2N
Formula Weight: 153.13
Melting Point: 39-44°
UN No.: 3439
MDL No.: MFCD07777154
Formula:
C₈H₅F₂N MW: 153,13 g/mol Melting Pt: 39…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD07777154 CAS Number: 847502-83-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...