3,5-Dichloro-4-hydroxybenzonitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
H50432.03
H50432.06
ALFAH50432.03EA
320
SGD
ALFAH50432.03
ALFAH50432.06
3,5-Dichloro-4-hydroxybenzonitrile ≥97%
3,5-Dichloro-4-hydroxybenzonitrile
Purity: 97+%
CAS No.: 1891-95-8
Molecular Formula: C7H3Cl2NO
Formula Weight: 188.01
Melting Point: 143-144°
UN No.: 3439
MDL No.: MFCD00002177
Formula:
C₇H₃Cl₂NO MW: 188,01 g/mol Melting Pt: 143…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002177 CAS Number: 1891-95-8 EINECS: 217-572-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...