2-Pyridineboronic acid dimethyl ester 95%
Supplier: Thermo Scientific
|
Warning
|
H53067.MD
H53067.03
ALFAH53067.MDEA
291
SGD
ALFAH53067.MD
ALFAH53067.03
2-Pyridineboronic acid dimethyl ester 95%
2-Pyridineboronic acid dimethyl ester
Purity: 95%
CAS No.: 136805-54-4
Molecular Formula: C7H10BNO2
Formula Weight: 150.97
MDL No.: MFCD03412109
Formula:
C₇H₁₀BNO₂ MW: 150,97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412109 CAS Number: 136805-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...