4-Butylbenzonitrile ≥98%
Supplier: Thermo Scientific
|
Warning
|
A19174.06
A19174.14
ALFAA19174.06EA
0
SGD
ALFAA19174.06
ALFAA19174.14
4-Butylbenzonitrile ≥98%
4-Butylbenzonitrile
Purity: 98%
CAS No.: 20651-73-4
Molecular Formula: C11H13N
Formula Weight: 159.23
Odour: Almond-like
Boiling Point: 130-132°/15mm
MDL No.: MFCD00052345
Formula:
C₁₁H₁₃N MW: 159,23 g/mol Boiling Pt: 130…132 °C (15 mmHg) Density: 0,924 Storage Temperature: Ambient |
MDL Number:
MFCD00052345 CAS Number: 20651-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...