Methyl-3,5-dibromo-4-methylbenzoate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 3,5-dibromo-4-methylbenzoate

B25348.06 B25348.14
ALFAB25348.06EA 0 SGD
ALFAB25348.06 ALFAB25348.14
Methyl-3,5-dibromo-4-methylbenzoate ≥98%
Methyl 3,5-dibromo-4-methylbenzoate

Purity: 98% CAS No.: 74896-66-5 Molecular Formula: C9H8Br2O2 Formula Weight: 307.98 Melting Point: 86-90° Beilstein Reference: 2580945

Formula: C₉H₈Br₂O₂
MW: 307.98 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00010729
CAS Number: 74896-66-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR