Methyl-4-bromo-2-(bromomethyl)benzoate 96%

Supplier: Thermo Scientific

Synonyms: Methyl 4-bromo-2-bromomethylbenzoate

H52350.MD H52350.03
ALFAH52350.MDEA 150 SGD
ALFAH52350.MD ALFAH52350.03
Methyl-4-bromo-2-(bromomethyl)benzoate 96%
Methyl 4-bromo-2-(bromomethyl)benzoate

Purity: 96% CAS No.: 78471-43-9 Molecular Formula: C9H8Br2O2 Formula Weight: 307.97 Melting Point: 76-78° MDL No.: MFCD04114319

Formula: C₉H₈Br₂O₂
MW: 307,97 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD04114319
CAS Number: 78471-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR