Methyl-2,6-dichloroisonicotinate ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 2,6-dichloropyridine-4-carboxylate, Methyl 2,6-dichloroisonicotinate, 2,6-Dichloro-4-pyridinecarboxylic acid methyl ester

A17468.03 A17468.06
ALFAA17468.03EA 0 SGD
ALFAA17468.03 ALFAA17468.06
Methyl-2,6-dichloroisonicotinate ≥97%
Methyl 2,6-dichloroisonicotinate

Purity: 97% CAS No.: 42521-09-5 Molecular Formula: C7H5Cl2NO2 Formula Weight: 206.03 Melting Point: 80-84° MDL No.: MFCD00044409

Formula: C₇H₅Cl₂NO₂
MW: 206,03 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00044409
CAS Number: 42521-09-5
EINECS: 255-868-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR