Methyl-2-aminoisonicotinate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

L19869.03 L19869.06
ALFAL19869.03EA 63.4 SGD
ALFAL19869.03 ALFAL19869.06
Methyl-2-aminoisonicotinate 97%
Methyl 2-aminoisonicotinate

Purity: 97% CAS No.: 6937-03-7 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: 145-149° Beilstein Reference: 128642

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD04039316
CAS Number: 6937-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR