(+)-DIOP ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4S,5S)-(+)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (S,S)-DIOP, (((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)
B21013.01
B21013.03
ALFAB21013.01EA
0
SGD
ALFAB21013.01
ALFAB21013.03
(+)-DIOP ≥98%
(+)-DIOP
Purity: 98%
CAS No.: 37002-48-5
Molecular Formula: C31H32O2P2
Formula Weight: 498.55
Storage Temperature: 2°C to 8°C
Melting Point: 88-90°
Beilstein Reference: 1630280
Optical Rotation: +12.5° (c=4.6 in benzene)
MDL No.: MFCD00009760
Formula:
C₃₁H₃₂O₂P₂ MW: 498,532 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD00009760 CAS Number: 37002-48-5 EINECS: 253-307-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...