3-Propylidenephthalide mixture E/Z 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Propylidene-1(3H)-isobenzofuranone, 3-Propylideneisobenzofuran-1(3H)-one
A13319.14
A13319.22
A13319.36
ALFAA13319.14EA
0
SGD
ALFAA13319.14
ALFAA13319.22
ALFAA13319.36
3-Propylidenephthalide mixture E/Z 96%
3-Propylidenephthalide
Purity: 96%
CAS No.: 17369-59-4
Molecular Formula: C11H10O2
Formula Weight: 174.20
Flash Point: 144°(291°F)
Refractive Index: 1.5870
Boiling Point: 101-103°/0.01m
Beilstein Reference: 4486
MDL No.: MFCD00051813
Formula:
C₁₁H₁₀O₂ MW: 174,20 g/mol Boiling Pt: 101…103 °C (0,01 mmHg) Density: 1,129 Flash Pt: 144 °C (291 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051813 CAS Number: 17369-59-4 EINECS: 241-402-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...