5-(3-Pyridyl)-1H-tetrazole ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 3-(2H-Tetrazol-5-yl)pyridine

B20492.03 B20492.06
ALFAB20492.03EA 0 SGD
ALFAB20492.03 ALFAB20492.06
5-(3-Pyridyl)-1H-tetrazole ≥98%
5-(3-Pyridyl)-1H-tetrazole

Purity: 98% CAS No.: 3250-74-6 Molecular Formula: C6H5N5 Formula Weight: 147.14 Odour: Odourless Melting Point: ca 240° dec. MDL No.: MFCD00040281

Formula: C₆H₅N₅
MW: 147,14 g/mol
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040281
CAS Number: 3250-74-6
EINECS: 426-810-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR