4-(3-Chlorophenoxy)piperidine ≥98%

Supplier: Thermo Scientific
Warning

H50956.03 H50956.MD
ALFAH50956.03EA 650 SGD
ALFAH50956.03 ALFAH50956.MD
4-(3-Chlorophenoxy)piperidine ≥98%
4-(3-Chlorophenoxy)piperidine

Purity: 98% CAS No.: 97840-40-9 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD08061087

Formula: C₁₁H₁₄ClNO
MW: 211,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061087
CAS Number: 97840-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR