3-(3-Chlorophenoxy)piperidine hydrochloride

Supplier: Thermo Scientific
Warning

H51180.MD H51180.03
ALFAH51180.MDEA 184 SGD
ALFAH51180.MD ALFAH51180.03
3-(3-Chlorophenoxy)piperidine hydrochloride
3-(3-Chlorophenoxy)piperidine hydrochloride

CAS No.: 1185301-51-2 Molecular Formula: C11H14ClNO.HCl Formula Weight: 248.15 MDL No.: MFCD11870724

Formula: C₁₁H₁₅Cl₂NO
MW: 248,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870724
CAS Number: 1185301-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR