4-(3-Methoxyphenoxy)piperidine ≥99%

Supplier: Thermo Scientific
Warning

H50964.03 H50964.MD
ALFAH50964.03EA 422 SGD
ALFAH50964.03 ALFAH50964.MD
4-(3-Methoxyphenoxy)piperidine ≥99%
4-(3-Methoxyphenoxy)piperidine

Purity: 99% CAS No.: 162402-37-1 Molecular Formula: C12H17NO2 Formula Weight: 207.27 MDL No.: MFCD08061092

Formula: C₁₂H₁₇NO₂
MW: 207,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061092
CAS Number: 162402-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR