2-(1-Piperazinyl)nicotinamide 95%
Supplier: Thermo Scientific
|
Warning
|
H50967.03
H50967.MD
ALFAH50967.03EA
430
SGD
ALFAH50967.03
ALFAH50967.MD
2-(1-Piperazinyl)nicotinamide 95%
2-(1-Piperazinyl)nicotinamide
Purity: 95%
CAS No.: 87394-64-7
Molecular Formula: C10H14N4O
Formula Weight: 206.25
MDL No.: MFCD08061093
Formula:
C₁₀H₁₄N₄O MW: 206,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061093 CAS Number: 87394-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...