1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt ≥97%

Supplier: Thermo Scientific
Warning

B23461.06 B23461.14
ALFAB23461.06EA 0 SGD
ALFAB23461.06 ALFAB23461.14
1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt ≥97%
1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt

Purity: 97% CAS No.: 96107-94-7 Molecular Formula: C4H5N4NaO2 Formula Weight: 164.11 Melting Point: ca 240° dec. UN No.: 1325 MDL No.: MFCD02683105

Formula: C₄H₅N₄NaO₂
MW: 164,11 g/mol
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02683105
CAS Number: 96107-94-7
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR