(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester ≥98%

Supplier: Thermo Fisher Scientific

L19650.03 L19650.06
ALFAL19650.03EA 0 SGD
ALFAL19650.03 ALFAL19650.06
(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester ≥98%
(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester

Purity: 98% CAS No.: 185427-48-9 Molecular Formula: C18H34B2O4 Formula Weight: 336.09 Flash Point: 110°(230°F) Refractive Index: 1.4560 Boiling Point: 102-104°/0.08mm Beilstein Reference: 7768251 MDL No.: MFCD03093898

Formula: C₁₈H₃₄B₂O₄
MW: 336.09 g/mol
Boiling Pt: 102…104 °C (0.08 mmHg)
Density: 0.954
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03093898
CAS Number: 185427-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR