(4-Chlorophenylthio)acetonitrile ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: [(4-Chlorophenyl)sulphanyl]acetonitrile

A12319.03 A12319.06
ALFAA12319.03EA 0 SGD
ALFAA12319.03 ALFAA12319.06
(4-Chlorophenylthio)acetonitrile ≥98%
(4-Chlorophenylthio)acetonitrile

Purity: 98% CAS No.: 18527-19-0 Molecular Formula: C8H6ClNS Formula Weight: 183.66 Melting Point: 79-81° MDL No.: MFCD00019832

Formula: C₈H₆ClNS
MW: 183,66 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00019832
CAS Number: 18527-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR