(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate
L14149.03
L14149.MD
ALFAL14149.03EA
0
SGD
ALFAL14149.03
ALFAL14149.MD
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Purity: 98+%
CAS No.: 39648-67-4
Molecular Formula: C20H13O4P
Formula Weight: 348.29
Melting Point:
Formula:
C₂₀H₁₃O₄P MW: 348,289 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010045 CAS Number: 39648-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...