2-Methoxy-6-methylpyridine ≥98%

Supplier: Thermo Scientific
Warning

H25772.03 H25772.06
ALFAH25772.03EA 0 SGD
ALFAH25772.03 ALFAH25772.06
2-Methoxy-6-methylpyridine ≥98%
2-Methoxy-6-methylpyridine

Purity: 98% CAS No.: 63071-03-4 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 49°(120°F) Refractive Index: 1.4995 Boiling Point: 156° UN No.: 1993 MDL No.: MFCD04112507

Formula: C₇H₉NO
MW: 123,16 g/mol
Boiling Pt: 156 °C
Density: 1,01
Flash Pt: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD04112507
CAS Number: 63071-03-4
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR