5-Phenyl-1-pentenylboronic acid 96%
Supplier: Thermo Scientific
Synonyms:
5-Phenyl-1-pentenylboronic acid pinacol ester
L19701.03
L19701.MD
ALFAL19701.03EA
498
SGD
ALFAL19701.03
ALFAL19701.MD
5-Phenyl-1-pentenylboronic acid 96%
5-Phenyl-1-pentenylboronic acid
Purity: 96%
CAS No.: 154820-97-0
Molecular Formula: C17H25BO2
Formula Weight: 272.20
Refractive Index: 1.4980
Beilstein Reference: 7295229
Formula:
C₁₇H₂₅BO₂ MW: 272,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788752 CAS Number: 154820-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...