4-Cyano-4'-octylbiphenyl ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Cyano-4'-n-octylbiphenyl, 4'-n-Octylbiphenyl-4-carbonitrile, 4'-n-Octyl-4-biphenylcarbonitrile, 8CB

H26318.MD H26318.03
ALFAH26318.MDEA 0 SGD
ALFAH26318.MD ALFAH26318.03
4-Cyano-4'-octylbiphenyl ≥99%
4-Cyano-4'-octylbiphenyl

Purity: 99% CAS No.: 52709-84-9 Molecular Formula: C21H25N Formula Weight: 291.43 Flash Point: 110°(230°F) Melting Point: 20-22° MDL No.: MFCD00075146

Formula: C₂₁H₂₅N
MW: 291.43 g/mol
Melting Pt: 20…22 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075146
CAS Number: 52709-84-9
EINECS: 258-120-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR