Isobutyl toluene-4-sulphonate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Isobutyl 4-methylbenzenesulphonate, Isobutyl p-toluenesulphonate
B20080.06
B20080.14
ALFAB20080.06EA
0
SGD
ALFAB20080.06
ALFAB20080.14
Isobutyl toluene-4-sulphonate ≥97%
Isobutyl toluene-4-sulphonate
Purity: 97%
CAS No.: 4873-56-7
Molecular Formula: C11H16O3S
Formula Weight: 228.28
Refractive Index: 1.5025
Formula:
C₁₁H₁₆O₃S MW: 228,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00026478 CAS Number: 4873-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...