4-Chloro-3-picoline hydrochloride ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Chloro-3-methylpyridine hydrochloride

L19558.03 L19558.MD
ALFAL19558.03EA 223 SGD
ALFAL19558.03 ALFAL19558.MD
4-Chloro-3-picoline hydrochloride ≥97%
4-Chloro-3-picoline hydrochloride

Purity: 97% CAS No.: 19524-08-4 Molecular Formula: C6H6ClN.HCl Formula Weight: 164.04 Odour: Odourless Melting Point: ca 185° Beilstein Reference: 107953 MDL No.: MFCD03092890

Formula: C₆H₇Cl₂N
MW: 164.04 g/mol
Melting Pt: ∼ 185 °C
Storage Temperature: Ambient
MDL Number: MFCD03092890
CAS Number: 19524-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR