2-Chloro-2-methylhexane ≥97%
Supplier: Thermo Scientific
|
Warning
|
B20183.06
B20183.14
ALFAB20183.06EA
0
SGD
ALFAB20183.06
ALFAB20183.14
2-Chloro-2-methylhexane ≥97%
2-Chloro-2-methylhexane
Purity: 97%
CAS No.: 4398-65-6
Molecular Formula: CH3(CH2)3CCl(CH3)2
Formula Weight: 134.65
UN No.: 1993
MDL No.: MFCD00060779
Formula:
C₇H₁₅Cl MW: 134,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060779 CAS Number: 4398-65-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...