Acetonitrile with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

NP461-20 461-4
BJAANP461-20EA 8620 SGD
BJAANP461-20 BJAA461-4
Acetonitrile with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile with 0,05% (v/v) formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C
Melting Pt: –46 °C
Density: 0,7765 g/cm3
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
EINECS: 208-759-1
UN: 1648
ADR: 3,II
Merck Index: 13,05212

Order Now

Specification Test Results

Formic Acid (v/v) 0.045 - 0.055%
UV Cutoff <189 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR