Acetonitrile with 0,1% (v/v) acetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

NP462-20 462-4
BJAANP462-20EA 5890 SGD
BJAANP462-20 BJAA462-4
Acetonitrile with 0,1% (v/v) acetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile with 0,1% (v/v) acetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 83 °C
Melting Pt: -46 °C
Density: 0,7770 g/cm3
Flash Pt: 19 °C
Storage Temperature: Ambient
EINECS: 208-759-1
UN: 1648
ADR: 3,II
Merck Index: 13,05212

Order Now

Specification Test Results

Acetic Acid (v/v) 0.09 - 0.11%
UV Cutoff <189 nm
Residue < 1 mg/L
Color (APHA) < 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR