Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON
Warning

NP452-20 452-4
BJAANP452-20EA 4470 SGD
BJAANP452-20 BJAA452-4
Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,1% (v/v) formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 214,4 °C (1013 hPa)
Melting Pt: 17 °C
Density: 0,997 g/cm3
Flash Pt: 99 °C
Storage Temperature: Ambient
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III
Merck Index: 13,09704

Order Now

Specification Test Results

Formic Acid (v/v) 0.09 - 0.11%
UV Cutoff <215 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR