Acetonitrile with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: BURDICK & JACKSON
|
Danger
|
NP441-20
441-4
BJAANP441-20EA
5690
SGD
BJAANP441-20
BJAA441-4
Acetonitrile with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile with 0,1% (v/v) Formic acid
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Boiling Pt:
82 °C (1,013 hPa) Melting Pt: –46 °C Density: 0,7822 g/cm³ (20 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
EINECS:
203-625-9 UN: 1648 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 13,09607 |
Specification Test Results
Formic Acid (v/v) | 0.09 - 0.11% |
UV Cutoff | <224 nm |
Residue | < 1 mg/L |
Color (APHA) | < 10 |
HPLC Gradient @ 215 nm | < 0.020 AU |
HPLC Gradient @ 254 nm | < 0.010 AU |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...